1-Acryloyl-3,5-bis(4-fluoro-3-nitrobenzylidene)azepan-4-one 98+%

Supplier: Ambeed
Warning

Synonyms: VLX1570

A150004-10MG A150004-5MG A150004-1MG
77162-740EA 54.59 USD
77162-740 77162-742 77611-390
1-Acryloyl-3,5-bis(4-fluoro-3-nitrobenzylidene)azepan-4-one 98+%
1-Acryloyl-3,5-bis(4-fluoro-3-nitrobenzylidene)azepan-4-one

Precaución: for Research Use Only

Formula: C₂₃H₁₇F₂N₃O₆
MW: 469.39 g/mol
MDL Number: MFCD28502165
CAS Number: 1431280-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR