1-(Trimethylsiloxy)cyclohexene 96% (by GC)

Supplier: Ambeed
Danger

Synonyms: (Cyclohex-1-en-1-yloxy)trimethylsilane

A149865-25G A149865-5G A149865-100G
77190-760EA 55 USD
77190-760 77190-762 77746-040
1-(Trimethylsiloxy)cyclohexene 96% (by GC)
1-(Trimethylsiloxy)cyclohexene

Precaución: for Research Use Only

Formula: C₉H₁₈OSi
MW: 170.33
MDL Number: MFCD00001541
CAS Number: 6651-36-1
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR