3-Bromo-7-iodoquinoline 95%

Supplier: Ambeed
Warning

A149679-100MG A149679-1G A149679-250MG A149679-5G
77190-752EA 35 USD
77190-752 77190-754 77190-756 77190-758
3-Bromo-7-iodoquinoline 95%
3-Bromo-7-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrIN
MDL Number: MFCD28679234
CAS Number: 1354223-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR