tert-Butyl (3R,5S,E)-7-(2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)-3,5-dihydroxyhept-6-enoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Buthyl Pitavastatin

A149477-1G A149477-250MG A149477-25G
77162-708EA 17.51 USD
77162-708 77162-710 77683-265
tert-Butyl (3R,5S,E)-7-(2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)-3,5-dihydroxyhept-6-enoate 97%
tert-Butyl (3R,5S,E)-7-(2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)-3,5-dihydroxyhept-6-enoate

Precaución: For research use only.

Formula: C₂₉H₃₂FNO₄
MW: 477.57 g/mol
MDL Number: MFCD12755990
CAS Number: 586966-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR