Z-D-Asp-OMe 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-D-Asp-OMe

A149207-10G A149207-1G A149207-250MG A149207-5G
77190-730EA 53 USD
77190-730 77190-732 77190-734 77190-736
Z-D-Asp-OMe 97%
Z-D-Asp-OMe

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₆
MDL Number: MFCD09842077
CAS Number: 47087-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR