1,5-Dibromo-2,6-dimethylnaphthalene 97%

Supplier: Ambeed
Warning

A149186-100MG A149186-1G A149186-250MG
77190-724EA 80.34 USD
77190-724 77190-726 77190-728
1,5-Dibromo-2,6-dimethylnaphthalene 97%
1,5-Dibromo-2,6-dimethylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀Br₂
MW: 314.01 g/mol
MDL Number: MFCD11053604
CAS Number: 20027-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR