2-(1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidine-4,5,6-triamine 98%
Supplier: Ambeed
|
Warning
|
A149052-1G
A149052-250MG
77162-688EA
63.86
USD
77162-688
77162-690
2-(1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidine-4,5,6-triamine 98%
2-(1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidine-4,5,6-triamine
Precaución: for Research Use Only
Formula:
C₁₇H₁₅FN₈ MW: 350.35 g/mol |
MDL Number:
MFCD22741544 CAS Number: 428854-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...