(Z)-6-(Pent-2-en-1-yl)tetrahydro-2H-pyran-2-one 97%

Supplier: Ambeed
Warning

A148565-1G A148565-25G A148565-5G
77190-704EA 22 USD
77190-704 77190-706 77190-708
(Z)-6-(Pent-2-en-1-yl)tetrahydro-2H-pyran-2-one 97%
(Z)-6-(Pent-2-en-1-yl)tetrahydro-2H-pyran-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MDL Number: MFCD00151946
CAS Number: 25524-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR