(S)-tert-Butyl 4-oxo-2-(thiazolidine-3-carbonyl)pyrrolidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A148473-100G
A148473-1G
A148473-25G
A148473-5G
77162-654EA
179.22
USD
77162-654
77162-656
77162-658
77162-660
(S)-tert-Butyl 4-oxo-2-(thiazolidine-3-carbonyl)pyrrolidine-1-carboxylate 95%
(S)-tert-Butyl 4-oxo-2-(thiazolidine-3-carbonyl)pyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₂₀N₂O₄S MW: 300.37 g/mol |
MDL Number:
MFCD22665915 CAS Number: 401564-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...