Ethyl 3-acetamido-5-chloro-2-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A148374-100MG A148374-1G A148374-250MG
77162-642EA 19 USD
77162-642 77162-644 77162-646
Ethyl 3-acetamido-5-chloro-2-hydroxybenzoate 98%
Ethyl 3-acetamido-5-chloro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClNO₄
MDL Number: MFCD28902469
CAS Number: 139329-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR