1-(3-Aminopropyl)-2-methylpiperidine 98%

Supplier: Ambeed
Danger

Synonyms: 1-(3-Aminopropyl)-2-methylhexahydropyridine, 2-Methyl-1-piperidinepropanamine, N-(3-Aminopropyl)-2-pipecoline, 3-(2-Methylpiperidin-1-yl)propan-1-amine, 1-(3-Aminopropyl)-2-methylpiperidine, 1-(3-Aminopropyl)-2-pipecoline

A148337-1G A148337-25G A148337-5G
77190-688EA 6.18 USD
77190-688 77190-690 77190-692
1-(3-Aminopropyl)-2-methylpiperidine 98%
1-(3-Aminopropyl)-2-methylpiperidine

Precaución: for Research Use Only

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
MDL Number: MFCD00006517
CAS Number: 25560-00-3
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR