Methyl 12-aminododecanoate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A148282-250MG
A148282-5G
A148282-25G
77162-636EA
44.29
USD
77162-636
77162-638
77683-263
Methyl 12-aminododecanoate hydrochloride 95%
Methyl 12-aminododecanoate hydrochloride
Precaución: For research use only.
Formula:
C₁₃H₂₈ClNO₂ |
MDL Number:
MFCD03788807 CAS Number: 4271-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...