4-(Ethoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A148089-250MG A148089-1G A148089-5G
77162-634EA 60.77 USD
77162-634 77683-258 77683-259
4-(Ethoxycarbonyl)benzoic acid 95%
4-(Ethoxycarbonyl)benzoic acid

Precaución: For research use only.

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD00496597
CAS Number: 713-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR