GCI1746 ≥98%

Supplier: Ambeed
Warning

A147108-10MG A147108-1MG A147108-25MG A147108-5MG
77162-592EA 47 USD
77162-592 77162-594 77162-596 77162-598
GCI1746 ≥98%
GCI1746

Precaución: for Research Use Only

Formula: C₃₄H₃₇N₅O₄
MDL Number: MFCD18782602
CAS Number: 910232-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR