1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Gatifloxacin hydrochloride

A146393-10G A146393-1MG A146393-5G A146393-1G A146393-250MG A146393-25G
77190-636EA 59.74 USD
77190-636 77190-638 77190-640 77746-027 77746-028 77746-029
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride 98%
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₃ClFN₃O₄
MW: 411.85 g/mol
MDL Number: MFCD09838901
CAS Number: 121577-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR