2-Amino-3-fluorobenzamide 98%

Supplier: Ambeed
Warning

A146355-1G A146355-250MG
77162-576EA 35 USD
77162-576 77162-578
2-Amino-3-fluorobenzamide 98%
2-Amino-3-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₇FN₂O
MDL Number: MFCD11109547
CAS Number: 187805-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR