1,5-Bis(acryloyloxy)pentane 97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A146184-100G A146184-10G A146184-1G A146184-25G A146184-500G
77190-620EA 72 USD
77190-620 77190-622 77190-624 77190-626 77190-628 77190-630
1,5-Bis(acryloyloxy)pentane 97% stabilized
1,5-Bis(acryloyloxy)pentane

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MDL Number: MFCD00078444
CAS Number: 36840-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR