tert-Butyl 4-(4-fluorophenyl)-3-oxopiperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A146059-100MG A146059-1G A146059-250MG
77162-570EA 19.57 USD
77162-570 77162-572 77162-574
tert-Butyl 4-(4-fluorophenyl)-3-oxopiperazine-1-carboxylate 95%
tert-Butyl 4-(4-fluorophenyl)-3-oxopiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉FN₂O₃
MW: 294.32 g/mol
MDL Number: MFCD16620878
CAS Number: 1284243-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR