4,5-Difluoro-o-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-1,2-phenylenediamine, 4,5-Difluorobenzene-1,2-diamine, 1,2-Diamino-4,5-difluorobenzene

A146055-10G A146055-1G A146055-25G A146055-5G A146055-100G A146055-250MG
77190-612EA 39.14 USD
77190-612 77190-614 77190-616 77190-618 77566-972 77566-974
4,5-Difluoro-o-phenylenediamine 98%
4,5-Difluoro-o-phenylenediamine

Precaución: For research use only.

Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
MDL Number: MFCD00061131
CAS Number: 76179-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR