4-Chloro-m-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-methylaniline, 5-Amino-2-chlorotoluene

A145839-10G A145839-1G A145839-5G
77190-602EA 14.42 USD
77190-602 77190-604 77746-019
4-Chloro-m-toluidine 98%
4-Chloro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6 g/mol
MDL Number: MFCD00066332
CAS Number: 7149-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR