Piperidine-3-carbonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A145824-250MG A145824-1G
77162-564EA 26.78 USD
77162-564 77592-728
Piperidine-3-carbonitrile hydrochloride 95%
Piperidine-3-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀N₂·ClH
MW: 146.618 g/mol
MDL Number: MFCD09037929
CAS Number: 1199773-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR