Sodium-3-chloro-2-hydroxypropanesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-hydroxypropanesulphonic acid Sodium Salt, Sodium 3-chloro-2-hydroxypropane-1-sulphonate

A145351-1000G A145351-100G A145351-10G A145351-25G A145351-500G A145351-5G
77162-540EA 98.88 USD
77162-540 77162-542 77162-544 77162-546 77162-548 77162-550
Sodium-3-chloro-2-hydroxypropanesulfonate 97%
Sodium-3-chloro-2-hydroxypropanesulfonate

Precaución: for Research Use Only

Formula: C₃H₇ClO₄S
MW: 196.58 g/mol
MDL Number: MFCD00013378
CAS Number: 126-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR