4-Nitrophenyl 5-chlorothiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A145245-1G A145245-250MG A145245-5G
77162-534EA 13 USD
77162-534 77162-536 77162-538
4-Nitrophenyl 5-chlorothiophene-2-carboxylate 97%
4-Nitrophenyl 5-chlorothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₆ClNO₄S
MDL Number: MFCD28099401
CAS Number: 1450877-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR