Methyl-2-chloro-4-thiazolecarboxylate 98%

Supplier: Ambeed
Warning

A145032-10G A145032-1G A145032-25G A145032-5G
77162-526EA 117.42 USD
77162-526 77162-528 77162-530 77162-532
Methyl-2-chloro-4-thiazolecarboxylate 98%
Methyl-2-chloro-4-thiazolecarboxylate

Precaución: for Research Use Only

Formula: C₅H₄ClNO₂S
MW: 177.6088 g/mol
MDL Number: MFCD06659909
CAS Number: 850429-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR