Methyl-3-hydroxy-1-methyl-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-hydroxy-1-methyl-1H-pyrrole-2-carboxylate

A144973-100MG A144973-1G A144973-250MG
77162-520EA 83 USD
77162-520 77162-522 77162-524
Methyl-3-hydroxy-1-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-3-hydroxy-1-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MDL Number: MFCD12924083
CAS Number: 113602-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR