3-(4-Methyl-2,5-dioxoimidazolidin-4-yl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methyl-2,5-dioxoimidazolidin-4-yl)propanoic acid

A144852-100MG A144852-1G
77162-516EA 63.86 USD
77162-516 77162-518
3-(4-Methyl-2,5-dioxoimidazolidin-4-yl)propionic acid 98%
3-(4-Methyl-2,5-dioxoimidazolidin-4-yl)propionic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₄
MW: 186.17 g/mol
MDL Number: MFCD08445781
CAS Number: 43189-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR