trans,trans-4-Propyl-4'-vinylbicyclohexyl 98%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-Propyl-4'-vinyl-1,1'-bi(cyclohexane)

A144262-25G A144262-5G A144262-1G
77190-550EA 78.28 USD
77190-550 77190-552 77746-013
trans,trans-4-Propyl-4'-vinylbicyclohexyl 98%
trans,trans-4-Propyl-4'-vinylbicyclohexyl

Precaución: for Research Use Only

Formula: C₁₇H₃₀
MW: 234.43 g/mol
MDL Number: MFCD11045059
CAS Number: 116020-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR