Tetrahydro-2H-pyran-4-ylhydrazine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (Tetrahydro-2H-pyran-4-yl)hydrazine hydrochloride

A144201-1G A144201-250MG A144201-5G
77190-544EA 21 USD
77190-544 77190-546 77190-548
Tetrahydro-2H-pyran-4-ylhydrazine hydrochloride 95%
Tetrahydro-2H-pyran-4-ylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₃ClN₂O
MDL Number: MFCD09744677
CAS Number: 194543-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR