Fmoc-D-isoGln-OH 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Glu-NH2

A143882-10G A143882-1G A143882-5G A143882-250MG
77162-482EA 56 USD
77162-482 77162-484 77683-240 77892-000
Fmoc-D-isoGln-OH 97%, Ambeed.Inc
Fmoc-D-isoGln-OH

Precaución: For research use only.

Formula: C₂₀H₂₀N₂O₅
MW: 368.4 g/mol
MDL Number: MFCD01861343
CAS Number: 292150-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR