Ethyl 5-oxoazepane-4-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A143668-100MG A143668-1G A143668-250MG A143668-5G
77162-470EA 38.11 USD
77162-470 77162-472 77162-474 77162-476
Ethyl 5-oxoazepane-4-carboxylate hydrochloride 95%
Ethyl 5-oxoazepane-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆ClNO₃
MW: 221.68 g/mol
MDL Number: MFCD17392855
CAS Number: 19673-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR