2-Chloro-5-iodopyridin-3-amine 97%

Supplier: Ambeed
Warning

A143657-1G A143657-250MG A143657-25G A143657-5G
77162-462EA 20 USD
77162-462 77162-464 77162-466 77162-468
2-Chloro-5-iodopyridin-3-amine 97%
2-Chloro-5-iodopyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MDL Number: MFCD04038465
CAS Number: 426463-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR