2-Chloro-3-methoxy-5-nitropyridine 98%

Supplier: Ambeed
Warning

A143548-100MG A143548-10G A143548-1G A143548-250MG
77162-454EA 20.6 USD
77162-454 77162-456 77162-458 77162-460
2-Chloro-3-methoxy-5-nitropyridine 98%
2-Chloro-3-methoxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₃
MW: 188.569 g/mol
MDL Number: MFCD06658397
CAS Number: 75711-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR