Methyl 2-aminopent-4-enoate hydrochloride 97%

Supplier: Ambeed
Warning

A143183-1G A143183-250MG A143183-5G
77162-430EA 119.48 USD
77162-430 77162-432 77162-434
Methyl 2-aminopent-4-enoate hydrochloride 97%
Methyl 2-aminopent-4-enoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·ClH
MW: 165.618 g/mol
MDL Number: MFCD25955960
CAS Number: 115289-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR