2',6'-Dichloro-3'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

A143075-1000G A143075-100G A143075-10G A143075-25G A143075-500G
77190-510EA 164.8 USD
77190-510 77190-512 77190-514 77190-516 77190-518
2',6'-Dichloro-3'-fluoroacetophenone 98%
2',6'-Dichloro-3'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
MDL Number: MFCD02093760
CAS Number: 290835-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR