5-Chloro-4-methylbenzene-1,3-diamine 98%

Supplier: Ambeed
Warning

A142887-100MG A142887-250MG
77162-422EA 45.32 USD
77162-422 77162-424
5-Chloro-4-methylbenzene-1,3-diamine 98%
5-Chloro-4-methylbenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.61 g/mol
MDL Number: MFCD11557587
CAS Number: 861519-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR