4-Amino-2-methylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-o-toluic acid, 4-Amino-2-methylbenzoic Acid

A142813-10G A142813-1G A142813-5G A142813-100G A142813-25G
77162-416EA 19.57 USD
77162-416 77162-418 77162-420 77683-233 77683-234
4-Amino-2-methylbenzoic acid 97%
4-Amino-2-methylbenzoic acid

Precaución: For research use only.

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD06656133
CAS Number: 2486-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR