2,4-Dimethylphenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dimethylphenyl)hydrazine hydrochloride, 2,4-Dimethylphenylhydrazine hydrochloride

A142677-10G A142677-25G A142677-5G
77190-500EA 12 USD
77190-500 77190-502 77190-504
2,4-Dimethylphenylhydrazinium chloride 97%
2,4-Dimethylphenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂
MDL Number: MFCD00013381
CAS Number: 60480-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR