4-Chloro-7-azaindole 98%
Supplier: Ambeed
|
Warning
|
A142462-100G
A142462-10G
A142462-1G
A142462-25G
A142462-5G
A142462-500G
77162-396EA
199.82
USD
77162-396
77162-398
77162-400
77162-402
77162-404
77683-231
4-Chloro-7-azaindole 98%
4-Chloro-7-azaindole
Precaución: For research use only.
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD08272232 CAS Number: 55052-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...