Methyl-2,2-dimethylacetoacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,2-dimethylacetoacetate

A142443-10G A142443-1G A142443-250MG A142443-25G A142443-5G
77162-386EA 67 USD
77162-386 77162-388 77162-390 77162-392 77162-394
Methyl-2,2-dimethylacetoacetate 95%
Methyl-2,2-dimethylacetoacetate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00026232
CAS Number: 38923-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR