4'-Borono-[1,1'-biphenyl]-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A142304-1G A142304-250MG A142304-5G
77190-490EA 150 USD
77190-490 77190-492 77190-494
4'-Borono-[1,1'-biphenyl]-4-carboxylic acid 97%
4'-Borono-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BO₄
MW: 242.03 g/mol
MDL Number: MFCD29077670
CAS Number: 872341-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR