5-Fluoro-2-methoxy-4-nitroaniline 95%
Supplier: Ambeed
|
Warning
|
A142204-10G
A142204-1G
A142204-5G
77162-382EA
216.3
USD
77162-382
77162-384
77683-230
5-Fluoro-2-methoxy-4-nitroaniline 95%
5-Fluoro-2-methoxy-4-nitroaniline
Precaución: For research use only.
Formula:
C₇H₇FN₂O₃ MW: 186.14 g/mol |
MDL Number:
MFCD24842657 CAS Number: 1435806-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...