(R)-2-Aminobut-3-enoic acid hydrochloride 95%

Supplier: Ambeed
Warning

A142094-100MG A142094-250MG A142094-50MG
77162-368EA 81.37 USD
77162-368 77162-370 77162-372
(R)-2-Aminobut-3-enoic acid hydrochloride 95%
(R)-2-Aminobut-3-enoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO₂
MW: 137.57 g/mol
MDL Number: MFCD01862356
CAS Number: 105763-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR