Diphenyl((phenylthio)methyl)phosphine oxide 95%

Supplier: Ambeed
Warning

A141786-100MG A141786-250MG A141786-50MG
77162-350EA 139.05 USD
77162-350 77162-352 77162-354
Diphenyl((phenylthio)methyl)phosphine oxide 95%
Diphenyl((phenylthio)methyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₉H₁₇OPS
MW: 324.37 g/mol
MDL Number: MFCD00482300
CAS Number: 31238-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR