2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

A141626-1G A141626-250MG A141626-5G
77162-332EA 107 USD
77162-332 77162-334 77891-987
2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester 98%
2-(Dimethoxymethyl)pyridine-5-boronic acid, pinacol ester
Formula: C₁₄H₂₂BNO₄
MDL Number: MFCD16038216
CAS Number: 1150632-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR