1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%

Supplier: Ambeed
Warning

Synonyms: SIPr.HCl, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium chloride, 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride

A141615-10G A141615-1G A141615-250MG A141615-25G A141615-5G A141615-100G
77190-454EA 53.56 USD
77190-454 77190-456 77190-458 77190-460 77190-462 77746-001
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride

Precaución: for Research Use Only

Formula: C₂₇H₃₉ClN₂
MW: 427.07 g/mol
MDL Number: MFCD07369796
CAS Number: 258278-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR