LY-2603618 ≥98%

Supplier: Ambeed
Warning

Synonyms: Rabusertib, (S)-1-(5-bromo-4-methyl-2-(morpholin-2-ylmethoxy)phenyl)-3-(5-methylpyrazin-2-yl)urea

A141550-10MG A141550-1MG A141550-5MG
77190-446EA 66.95 USD
77190-446 77190-448 77190-450
LY-2603618 ≥98%
LY-2603618

Precaución: for Research Use Only

Formula: C₁₈H₂₂BrN₅O₃
MW: 436.3 g/mol
MDL Number: MFCD18633253
CAS Number: 911222-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR