3-Nitrophthalic anhydride 96%

Supplier: Ambeed
Warning

A141291-100G A141291-10G A141291-25G A141291-500G
77190-432EA 12 USD
77190-432 77190-434 77190-436 77190-438
3-Nitrophthalic anhydride 96%
3-Nitrophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₃NO₅
MDL Number: MFCD00005921
CAS Number: 641-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR