3-Phenyl-1H-indole 97%

Supplier: Ambeed
Warning

A141175-100MG A141175-250MG A141175-1G
77190-428EA 28 USD
77190-428 77190-430 77566-844
3-Phenyl-1H-indole 97%
3-Phenyl-1H-indole

Precaución: For research use only.

Formula: C₁₄H₁₁N
MDL Number: MFCD00033474
CAS Number: 1504-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR