1,2-Dihydro-3H-1,2,4-triazol-3-one 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-1,2,4-Triazol-3(2H)-one
A141044-100G
A141044-10G
A141044-25G
A141044-500G
A141044-5G
77190-418EA
75.19
USD
77190-418
77190-420
77190-422
77190-424
77190-426
1,2-Dihydro-3H-1,2,4-triazol-3-one 97%
1,2-Dihydro-3H-1,2,4-triazol-3-one
Precaución: for Research Use Only
Formula:
C₂H₃N₃O MW: 85.065 g/mol |
MDL Number:
MFCD01311031 CAS Number: 930-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...