1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride 97%

Supplier: Ambeed
Warning

Synonyms: O-(2,4-Dichlorobenzyl)hydroxylamine hydrochloride, O-[(2,4-Dichlorophenyl)methyl]hydroxylamine hydrochloride

A140869-1G A140869-250MG A140869-25G A140869-5G
77190-410EA 16.48 USD
77190-410 77190-412 77190-414 77190-416
1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride 97%
1-[(Ammoniooxy)methyl]-2,4-dichlorobenzene chloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₃NO
MW: 228.5000 g/mol
MDL Number: MFCD01114586
CAS Number: 51572-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR